Stampa…
2-Cyclohexylethylamine hydrochloride 95%
Fornitore: Apollo Scientific
Synonyms: (2-Aminoethyl)cyclohexane hydrochloride, 2-Cyclohexylethylamine hydrochloride
OR925622-1G OR925622-5G
APOSOR925622-1GEA 150 EUR
APOSOR925622-1G APOSOR925622-5G
2-Cyclohexylethylamine hydrochloride 95%
2-Cyclohexylethylamine hydrochloride
Formula: C₈H₁₈ClN
Peso molecolare: 163.688 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01734731
CAS Number: 5471-55-6