Stampa…

6-Acetyl-2-Benzoxazolinone 95%

Fornitore: Apollo Scientific

Synonyms: 6-Acetylbenzo[d]oxazol-2(3H)-one

OR925460-1G OR925460-5G
APOSOR925460-1GEA 139 EUR
APOSOR925460-1G APOSOR925460-5G
6-Acetyl-2-Benzoxazolinone 95%
6-Acetyl-2-Benzoxazolinone
Formula: C₉H₇NO₃
Peso molecolare: 177,16 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01664312
CAS Number: 54903-09-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR