4-Methylpyrocatechol 95%
Fornitore: Apollo Scientific
Formula:
C₇H₈O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002205 CAS Number: 452-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...