Stampa…

6-Methyl-1,2,3,4-tetrahydroquinoline 97%

Fornitore: Apollo Scientific

Synonyms: 1,2,3,4-Tetrahydro-6-methylquinoline

OR925199-25G OR925199-5G
APOSOR925199-25GEA 448 EUR
APOSOR925199-25G APOSOR925199-5G
6-Methyl-1,2,3,4-tetrahydroquinoline 97%
6-Methyl-1,2,3,4-tetrahydroquinoline
Formula: C₁₀H₁₃N
Peso molecolare: 147,22 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00023887
CAS Number: 91-61-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR