Tetrahydrofurfuryl acrylate 95%
Fornitore: Apollo Scientific
Formula:
C₈H₁₂O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00014516 CAS Number: 2399-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...