Ethyl benzenesulphonate 95%
Fornitore: Apollo Scientific
Formula:
C₈H₁₀O₃S Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00025033 CAS Number: 515-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...