Stampa…

Ethyl-3-aminobutanoate 98%

Fornitore: Apollo Scientific

Synonyms: Ethyl 3-aminobutyrate

OR924945-100G OR924945-25G OR924945-5G
APOSOR924945-100GEA 931 EUR
APOSOR924945-100G APOSOR924945-25G APOSOR924945-5G
Ethyl-3-aminobutanoate 98%
Ethyl-3-aminobutanoate
Formula: C₆H₁₃NO₂
Peso molecolare: 131,18 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008086
CAS Number: 5303-65-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR