Stampa…

1-Ethoxynaphthalene 95%

Fornitore: Apollo Scientific

Synonyms: Ethyl 1-naphthyl ether, 1-Ehoxynaphthalene

OR923772-25G OR923772-5G
APOSOR923772-25GEA 865 EUR
APOSOR923772-25G APOSOR923772-5G
1-Ethoxynaphthalene 95%
1-Ethoxynaphthalene
Formula: C₁₂H₁₂O
Peso molecolare: 172,23 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003928
CAS Number: 5328-01-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR