Stampa…

2-Cyclopentylcyclopentanone 95%

Fornitore: Apollo Scientific

Synonyms: [1,1'-Bi(cyclopentan)]-2-one

OR923715-100G OR923715-25G
APOSOR923715-100GEA 328 EUR
APOSOR923715-100G APOSOR923715-25G
2-Cyclopentylcyclopentanone 95%
2-Cyclopentylcyclopentanone
Formula: C₁₀H₁₆O
Peso molecolare: 152,24 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00051419
CAS Number: 4884-24-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR