Stampa…

(S)-(-)-3-Cyclohexene-1-carboxylic acid 95%

Fornitore: Apollo Scientific

Synonyms: (S)-Cyclohex-3-enecarboxylic acid, (S)-(-)-1,2,3,6-Tetrahydrobenzoic acid

OR923698-10G OR923698-25G OR923698-5G
APOSOR923698-10GEA 248 EUR
APOSOR923698-10G APOSOR923698-25G APOSOR923698-5G
(S)-(-)-3-Cyclohexene-1-carboxylic acid 95%
(S)-(-)-3-Cyclohexene-1-carboxylic acid
Formula: C₇H₁₀O₂
Peso molecolare: 126,16 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08275437
CAS Number: 5708-19-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR