Stampa…

4,4'-Bicyclohexanol 95%

Fornitore: Apollo Scientific

Synonyms: 1,1'-Bicyclohexyl-4,4'-diol, [1,1'-Bi(cyclohexane)]-4,4'-diol, 4,4'-Dihydroxydicyclohexyl

OR923611-100G OR923611-25G OR923611-5G
APOSOR923611-100GEA 78.6 EUR
APOSOR923611-100G APOSOR923611-25G APOSOR923611-5G
4,4'-Bicyclohexanol 95%
4,4'-Bicyclohexanol
Formula: C₁₂H₂₂O₂
Peso molecolare: 198,31 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012056
CAS Number: 20601-38-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR