2-Cyclohexylcyclohexanol 95%
Fornitore: Apollo Scientific
Synonyms:
[1,1'-Bi(cyclohexan)]-2-ol
OR923305-25G
OR923305-5G
APOSOR923305-25GEA
437
EUR
APOSOR923305-25G
APOSOR923305-5G
2-Cyclohexylcyclohexanol 95%
2-Cyclohexylcyclohexanol
Formula:
C₁₂H₂₂O Peso molecolare: 182,31 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00021278 CAS Number: 6531-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...