Stampa…

Ethynyl-p-tolyl sulphone 95%

Fornitore: Apollo Scientific

Synonyms: Tosylacetylene, Ethynyl p-Tolyl sulphone, p-Toluenesulphonylacetylene

OR923266-1G OR923266-250MG
APOSOR923266-1GEA 423 EUR
APOSOR923266-1G APOSOR923266-250MG
Ethynyl-p-tolyl sulphone 95%
Ethynyl-p-tolyl sulphone
Formula: C₉H₈O₂S
Peso molecolare: 180,22 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00191647
CAS Number: 13894-21-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR