Stampa…

N,N-Diethylbenzamide 95%

Fornitore: Apollo Scientific

OR922828-25G OR922828-100G
APOSOR922828-25GEA 88.7 EUR
APOSOR922828-25G APOSOR922828-100G
N,N-Diethylbenzamide 95%
N,N-Diethylbenzamide
Formula: C₁₁H₁₅NO
Peso molecolare: 177,25 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00026726
CAS Number: 1696-17-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR