Stampa…

2-Chloro-6-ethoxypyridine 97%

Fornitore: Apollo Scientific

OR922826-1G OR922826-25G OR922826-5G
APOSOR922826-1GEA 125 EUR
APOSOR922826-1G APOSOR922826-25G APOSOR922826-5G
2-Chloro-6-ethoxypyridine 97%
2-Chloro-6-ethoxypyridine
Formula: C₇H₈ClNO
Peso molecolare: 157,6 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00023465
CAS Number: 42144-78-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR