Stampa…

Dimethyl isophthalate 95%

Fornitore: Apollo Scientific

Synonyms: Isophthalic acid dimethyl ester

OR922564-100G OR922564-1KG OR922564-500G
APOSOR922564-100GEA 51 EUR
APOSOR922564-100G APOSOR922564-1KG APOSOR922564-500G
Dimethyl isophthalate 95%
Dimethyl isophthalate
Formula: C₁₀H₁₀O₄
Peso molecolare: 194,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008433
CAS Number: 1459-93-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR