Pentyl nitrate 95%
Fornitore: Apollo Scientific
Synonyms:
Pentyl nitrate, Amyl nitrate
OR922460-1G
OR922460-25G
OR922460-5G
APOSOR922460-1GEA
147
EUR
APOSOR922460-1G
APOSOR922460-25G
APOSOR922460-5G
Pentyl nitrate 95%
Pentyl nitrate
Formula:
C₅H₁₁NO₃ Peso molecolare: 133,15 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00059177 CAS Number: 1002-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...