Stampa…

1-Acetylpiperidine 95%

Fornitore: Apollo Scientific

Synonyms: 1-(Piperidin-1-yl)ethanone

OR921648-1G OR921648-25G OR921648-5G
APOSOR921648-1GEA 49.5 EUR
APOSOR921648-1G APOSOR921648-25G APOSOR921648-5G
1-Acetylpiperidine 95%
1-Acetylpiperidine
Formula: C₇H₁₃NO
Peso molecolare: 127,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006486
CAS Number: 618-42-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR