2'-Ethoxyacetanilide 95%
Fornitore: Apollo Scientific
Synonyms:
N-Acetyl-o-phenetidine
OR920903-25G
OR920903-5G
OR920903-100G
APOSOR920903-25GEA
299
EUR
APOSOR920903-25G
APOSOR920903-5G
APOSOR920903-100G
2'-Ethoxyacetanilide 95%
2'-Ethoxyacetanilide
Formula:
C₁₀H₁₃NO₂ Peso molecolare: 179,22 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00059228 CAS Number: 581-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...