Hexanamide 95%
Fornitore: Apollo Scientific
Synonyms:
Capronamide
OR920698-25G
OR920698-5G
APOSOR920698-25GEA
166
EUR
APOSOR920698-25G
APOSOR920698-5G
Hexanamide 95%
Hexanamide
Formula:
C₆H₁₃NO Peso molecolare: 115,18 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008046 CAS Number: 628-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...