2-(3-Chloropropoxy)tetrahydro-2H-pyran 95%
Fornitore: Apollo Scientific
OR920428-25G
OR920428-5G
APOSOR920428-25GEA
343
EUR
APOSOR920428-25G
APOSOR920428-5G
2-(3-Chloropropoxy)tetrahydro-2H-pyran 95%
2-(3-Chloropropoxy)tetrahydro-2H-pyran
Formula:
C₈H₁₅ClO₂ Peso molecolare: 178,66 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00023846 CAS Number: 42330-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...