Stampa…
Trimethylpropylammonium bromide 95%
Fornitore: Apollo Scientific
Synonyms: N,N,N-Trimethylpropan-1-aminium bromide
OR919812-1G OR919812-25G OR919812-5G
APOSOR919812-1GEA 53.9 EUR
APOSOR919812-1G APOSOR919812-25G APOSOR919812-5G
Trimethylpropylammonium bromide 95%
Trimethylpropylammonium bromide
Formula: C₆H₁₆BrN
Peso molecolare: 182.11 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06227832
CAS Number: 2650-50-2