Stampa…

2,2'-Dipyridylamine 99%

Fornitore: Apollo Scientific

Synonyms: 2,2'-Iminodipyridine, Di(pyridin-2-yl)amine, Chlorpheniramine Related Compound B

OR919705-25G OR919705-5G
APOSOR919705-25GEA 423 EUR
APOSOR919705-25G APOSOR919705-5G
2,2'-Dipyridylamine 99%
2,2'-Dipyridylamine
Formula: C₁₀H₉N₃
Peso molecolare: 171.2 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006247
CAS Number: 1202-34-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR