Stampa…

4-(Dimethylamino)-1-butanol 98%

Fornitore: Apollo Scientific

Synonyms: 4-(Dimethylamino)butan-1-ol, 4-Dimethylamino-1-butanol

OR919427-100G OR919427-25G OR919427-5G
APOSOR919427-100GEA 807 EUR
APOSOR919427-100G APOSOR919427-25G APOSOR919427-5G
4-(Dimethylamino)-1-butanol 98%
4-(Dimethylamino)-1-butanol
Formula: C₆H₁₅NO
Peso molecolare: 117,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00020624
CAS Number: 13330-96-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR