Stampa…

5-Norbornene-2-exo,3-exo-dimethanol 97%

Fornitore: Apollo Scientific

Synonyms: 5-exo,6-exo-Bis(hydroxymethyl)-2-norbornene, Bicyclo[2.2.1]hept-5-ene-2-exo,3-exo-dimethanol

OR919253-1G OR919253-5G
APOSOR919253-1GEA 183 EUR
APOSOR919253-1G APOSOR919253-5G
5-Norbornene-2-exo,3-exo-dimethanol 97%
5-Norbornene-2-exo,3-exo-dimethanol
Formula: C₉H₁₄O₂
Peso molecolare: 154,21 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00799568
CAS Number: 699-95-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR