Stampa…

2-(Chloromethyl)-4-methylquinazoline 95%

Fornitore: Apollo Scientific

OR918316-10G OR918316-1G OR918316-5G
APOSOR918316-10GEA 105 EUR
APOSOR918316-10G APOSOR918316-1G APOSOR918316-5G
2-(Chloromethyl)-4-methylquinazoline 95%
2-(Chloromethyl)-4-methylquinazoline
Formula: C₁₀H₉ClN₂
Peso molecolare: 192,65 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09807547
CAS Number: 109113-72-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR