Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl) 95%
Fornitore: Apollo Scientific
Synonyms:
H-Gly-OIpr·HCl, Isopropyl 2-aminoacetate hydrochloride
OR918059-25G
OR918059-5G
APOSOR918059-25GEA
154
EUR
APOSOR918059-25G
APOSOR918059-5G
Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl) 95%
Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl)
Formula:
C₅H₁₂ClNO₂ Peso molecolare: 153,61 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00151826 CAS Number: 14019-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...