5-Oxohexanoic acid 98%
Fornitore: Apollo Scientific
OR917515-100G
OR917515-25G
APOSOR917515-100GEA
265
EUR
APOSOR917515-100G
APOSOR917515-25G
5-Oxohexanoic acid 98%
5-Oxohexanoic acid
Formula:
C₆H₁₀O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004412 CAS Number: 3128-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...