N-Ethyl-2-pyrrolidin-1-ylethanamine 95%
Fornitore: Apollo Scientific
Synonyms:
N-Ethyl-2-(1-pyrrolidyl)ethanamine
OR916855-1G
OR916855-250MG
APOSOR916855-1GEA
125
EUR
APOSOR916855-1G
APOSOR916855-250MG
N-Ethyl-2-pyrrolidin-1-ylethanamine 95%
N-Ethyl-2-pyrrolidin-1-ylethanamine
Formula:
C₈H₁₈N₂ Peso molecolare: 142,25 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08361605 CAS Number: 138356-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...