4-(1H-Pyrazol-1-ylmethyl)benzaldehyde 95%
Fornitore: Apollo Scientific
OR916849-1G
OR916849-250MG
APOSOR916849-1GEA
401
EUR
APOSOR916849-1G
APOSOR916849-250MG
4-(1H-Pyrazol-1-ylmethyl)benzaldehyde 95%
4-(1H-Pyrazol-1-ylmethyl)benzaldehyde
Formula:
C₁₁H₁₀N₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08059835 CAS Number: 887922-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...