Stampa…

7-Hydroxy-2-tetralone 95%

Fornitore: Apollo Scientific

Synonyms: 7-Hydroxy-3,4-dihydro-1H-naphthalen-2-one

OR916604-1G OR916604-250MG
APOSOR916604-1GEA 379 EUR
APOSOR916604-1G APOSOR916604-250MG
7-Hydroxy-2-tetralone 95%
7-Hydroxy-2-tetralone
Formula: C₁₀H₁₀O₂
Peso molecolare: 162,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04038657
CAS Number: 37827-68-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR