Stampa…

4-Cyanophenylacetic acid 98%

Fornitore: Apollo Scientific

OR914390-1G OR914390-25G
APOSOR914390-1GEA 24.4 EUR
APOSOR914390-1G APOSOR914390-25G
4-Cyanophenylacetic acid 98%
4-Cyanophenylacetic acid
Formula: C₉H₇NO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06798066
CAS Number: 5462-71-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR