Stampa…

1,2,3,4-Tetrahydropyrrolo[1,2-a]pyrazine 95%

Fornitore: Apollo Scientific

OR914166-1G OR914166-250MG
APOSOR914166-1GEA 597 EUR
APOSOR914166-1G APOSOR914166-250MG
1,2,3,4-Tetrahydropyrrolo[1,2-a]pyrazine 95%
1,2,3,4-Tetrahydropyrrolo[1,2-a]pyrazine
Formula: C₇H₁₀N₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00296976
CAS Number: 71257-38-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR