Stampa…

(R)-1-(pyridin-4-yl)ethanol 97%

Fornitore: Apollo Scientific

Synonyms: (R)-alpha-Methyl-4-pyridinemethanol, (R)-1-(4-Pyridyl)ethanol, (R)-4-(1-Hydroxyethyl)pyridine, (R)-1-(Pyridin-4-yl)ethanol

OR913596-1G OR913596-250MG
APOSOR913596-1GEA 104 EUR
APOSOR913596-1G APOSOR913596-250MG
(R)-1-(pyridin-4-yl)ethanol 97%
(R)-1-(pyridin-4-yl)ethanol
Formula: C₇H₉NO
Peso molecolare: 123,155 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00077865
CAS Number: 27854-88-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR