Ethyl-(S)-4-Chloro-3-hydroxybutyrate 97%
Fornitore: Apollo Scientific
Synonyms:
(S)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester, Ethyl (S)-4-chloro-3-hydroxybutanoate
OR912718-100G
OR912718-10G
OR912718-25G
APOSOR912718-100GEA
94.5
EUR
APOSOR912718-100G
APOSOR912718-10G
APOSOR912718-25G
Ethyl-(S)-4-Chloro-3-hydroxybutyrate 97%
Ethyl-(S)-4-Chloro-3-hydroxybutyrate
Formula:
C₆H₁₁ClO₃ Peso molecolare: 166,60 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00211241 CAS Number: 86728-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...