Stampa…

3-Chloro-4'-methoxypropiophenone 95%

Fornitore: Apollo Scientific

Synonyms: 3-Chloro-1-(4-methoxyphenyl)propan-1-one

OR912485-1G OR912485-5G
APOSOR912485-1GEA 109 EUR
APOSOR912485-1G APOSOR912485-5G
3-Chloro-4'-methoxypropiophenone 95%
3-Chloro-4'-methoxypropiophenone
Formula: C₁₀H₁₁ClO₂
Peso molecolare: 198,65 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00236602
CAS Number: 35999-20-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR