Stampa…

Methyl tetrahydrofuran-2-carboxylate 97%

Fornitore: Apollo Scientific

Synonyms: 2-Tetrahydrofuroic Acid Methyl Ester, Tetrahydrofuran-2-carboxylic Acid Methyl Ester

OR912406-100G OR912406-500G
APOSOR912406-100GEA 168 EUR
APOSOR912406-100G APOSOR912406-500G
Methyl tetrahydrofuran-2-carboxylate 97%
Methyl tetrahydrofuran-2-carboxylate
Formula: C₆H₁₀O₃
Peso molecolare: 130,14 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00154541
CAS Number: 37443-42-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR