Stampa…
4-(Methylthio)-m-cresol 98%
Fornitore: Apollo Scientific
Synonyms: 3-Methyl-4-(methylthio)phenol
OR912189-100G OR912189-25G
APOSOR912189-100GEA 350 EUR
APOSOR912189-100G APOSOR912189-25G
4-(Methylthio)-m-cresol 98%
4-(Methylthio)-m-cresol
Formula: C₈H₁₀OS
Peso molecolare: 154,23 g/mol
MDL Number: MFCD00045773
CAS Number: 3120-74-9