Sodium dimethyldithiocarbamate 95%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
Dimethyldithiocarbamic acid sodium salt
Formula:
C₃H₇NS₂ Peso molecolare: 143.20 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00044839 CAS Number: 128-04-1 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...