Stampa…

2-Isopropyl-5-methyl-4-nitrosophenol 95%

Fornitore: Apollo Scientific

OR911954-1G OR911954-5G
APOSOR911954-1GEA 263 EUR
APOSOR911954-1G APOSOR911954-5G
2-Isopropyl-5-methyl-4-nitrosophenol 95%
2-Isopropyl-5-methyl-4-nitrosophenol
Formula: C₁₀H₁₃NO₂
Temperatura di stoccaggio: Ambiente
CAS Number: 2364-54-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR