Stampa…

3',5'-Dichloroacetophenone 95%

Fornitore: Apollo Scientific

Synonyms: 1-(3,5-Dichlorophenyl)ethanone

OR911946-10G OR911946-1G OR911946-5G
APOSOR911946-10GEA 94.5 EUR
APOSOR911946-10G APOSOR911946-1G APOSOR911946-5G
3',5'-Dichloroacetophenone 95%
3',5'-Dichloroacetophenone
Formula: C₈H₆Cl₂O
Peso molecolare: 189,04 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00045189
CAS Number: 14401-72-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR