7-Chloroisatin 97%
Fornitore: Apollo Scientific
Synonyms:
7-Chloroindoline-2,3-dione, 7-Chloro-1H-indole-2,3-dione, 7-Cloroindoline-2,3-dione
OR911682-100G
OR911682-25G
OR911682-5G
APOSOR911682-100GEA
539
EUR
APOSOR911682-100G
APOSOR911682-25G
APOSOR911682-5G
7-Chloroisatin 97%
7-Chloroisatin
Formula:
C₈H₄ClNO₂ Peso molecolare: 181,58 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00022796 CAS Number: 7477-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...