7-Methylquinoline 97%
Fornitore: Apollo Scientific
Formula:
C₁₀H₉N |
MDL Number:
MFCD00006805 CAS Number: 612-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...