2-Hydroxyquinoline-4-carboxylic acid 98%
Fornitore: Apollo Scientific
Synonyms:
2-Hydroxycinchoninic Acid
OR911510-100G
OR911510-500G
APOSOR911510-100GEA
104
EUR
APOSOR911510-100G
APOSOR911510-500G
2-Hydroxyquinoline-4-carboxylic acid 98%
2-Hydroxyquinoline-4-carboxylic acid
Formula:
C₁₀H₇NO₃ Peso molecolare: 189,17 g/mol Punto di fusione: 300 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00023942 CAS Number: 15733-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...