Stampa…

2-Furaldehyde diethyl acetal 95%

Fornitore: Apollo Scientific

Synonyms: 2-(Diethoxymethyl)furan, Furfural Diethyl Acetal

OR911440-1G OR911440-5G
APOSOR911440-1GEA 132 EUR
APOSOR911440-1G APOSOR911440-5G
2-Furaldehyde diethyl acetal 95%
2-Furaldehyde diethyl acetal
Formula: C₉H₁₄O₃
Peso molecolare: 170,21 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003245
CAS Number: 13529-27-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR