4-Butoxybenzoic acid 98%
Fornitore: Apollo Scientific
Formula:
C₁₁H₁₄O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002546 CAS Number: 1498-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...