Stampa…

N-Methylpiperidin-3-amine dihydrochloride 97%

Fornitore: Apollo Scientific

Synonyms: 3-(Methylamino)piperidine dihydrochloride

OR910998-100G OR910998-25G
APOSOR910998-100GEA 437 EUR
APOSOR910998-100G APOSOR910998-25G
N-Methylpiperidin-3-amine dihydrochloride 97%
N-Methylpiperidin-3-amine dihydrochloride
Formula: C₆H₁₆Cl₂N₂
Peso molecolare: 187,1100 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00673143
CAS Number: 127294-77-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR