Stampa…

Methyl-3-furoate 98%

Fornitore: Apollo Scientific

Synonyms: Methyl furan-3-carboxylate

OR910096-100G OR910096-25G OR910096-5G
APOSOR910096-100GEA 807 EUR
APOSOR910096-100G APOSOR910096-25G APOSOR910096-5G
Methyl-3-furoate 98%
Methyl-3-furoate
Formula: C₆H₆O₃
Peso molecolare: 126,11 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06203671
CAS Number: 13129-23-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR