Stampa…

2-Methylbenzoylacetonitrile 98%

Fornitore: Apollo Scientific

Synonyms: 3-(2-Methylphenyl)-3-oxopropanenitrile, 3-Oxo-3-(o-tolyl)propanenitrile

OR909663-10G OR909663-1G OR909663-5G
APOSOR909663-10GEA 757 EUR
APOSOR909663-10G APOSOR909663-1G APOSOR909663-5G
2-Methylbenzoylacetonitrile 98%
2-Methylbenzoylacetonitrile
Formula: C₁₀H₉NO
Peso molecolare: 159,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02260802
CAS Number: 35276-81-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR