5-Bromo-2-methylbenzaldehyde 97%
Fornitore: Apollo Scientific
OR909370-10G
OR909370-1G
OR909370-5G
APOSOR909370-10GEA
502
EUR
APOSOR909370-10G
APOSOR909370-1G
APOSOR909370-5G
5-Bromo-2-methylbenzaldehyde 97%
5-Bromo-2-methylbenzaldehyde
Formula:
C₈H₇BrO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD05662390 CAS Number: 90050-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...